Learn More
1-Piperazinecarboxaldehyde, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 388491000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Piperazinecarboxaldehyde | |
88.0 | |
1.1000g/mL | |
102°C | |
88% min. (GC) | |
C5H11N2O | |
MFCD00005963 | |
1.1 | |
MSSDTZLYNMFTKN-UHFFFAOYSA-O | |
piperazine-1-carbaldehyde | |
82191 | |
90% |
7755-92-2 | |
100.0 | |
105.0°C to 107.0°C (1.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5097 to 1.5117 | |
100g | |
1-formylpiperazine, 1-piperazinecarboxaldehyde, 1-piperazinecarbaldehyde, formylpiperazine, n-formylpiperazine, 1-formyl piperazine, piperazinecarbaldehyde, l-formylpiperazine, 4-formylpiperazine, n-formyl piperazine | |
O=CN1CC[NH2+]CC1 | |
115.16 | |
114.15 |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 231-813-
RUO â Research Use Only