missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenylvinylboronic acid pinacol ester, 95%

Manufacturer:  Alfa Aesar™ H53216MD

 View more versions of this product

Catalog No. AAH53216MD



Specifications

Specifications

1-Phenylvinylboronic acid pinacol ester
250mg
C14H19BO2
4,4,5,5-tetramethyl-2-1-phenylvinyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl, 1-phenylvinylboronic acid pinacolester, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C(=C)C2=CC=CC=C2
230.114
230.11
40°C to 42°C
143825-84-7
MFCD06659923
RMGBWPMWUZSIMH-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
11031671
95%

Chemical Identifiers

143825-84-7
230.114
RMGBWPMWUZSIMH-UHFFFAOYSA-N
11031671
B1(OC(C(O1)(C)C)(C)C)C(=C)C2=CC=CC=C2
C14H19BO2
MFCD06659923
4,4,5,5-tetramethyl-2-1-phenylvinyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl, 1-phenylvinylboronic acid pinacolester, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
SDS Documents