Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Phenylsulfonyl)pyrrole, 98%

$47.57 - $648.04

Chemical Identifiers

Chemical Name or Material 1- (Phenylsulfonyl)pyrrole
CAS 16851-82-4
Molecular Formula C10H9NO2S
Formula Weight 207.25
MDL Number MFCD00067739
Beilstein 1619427
Synonym 1-(Benzenesulfonyl)pyrrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1159903

 
alfa aesar™
A1159903
1g Each for $47.57

AAA1159906

 
alfa aesar™
A1159906
5g Each for $166.24

AAA1159914

 
alfa aesar™
A1159914
25g Each for $648.04
Description & Specifications

Specifications

Formula Weight 207.25
Chemical Name or Material 1- (Phenylsulfonyl)pyrrole
Assay 98%
Molecular Formula C10H9NO2S
MDL Number MFCD00067739
CAS 16851-82-4
Beilstein 1619427
Synonym 1-(Benzenesulfonyl)pyrrole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 87° to 90°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.