Please login to your online account to display your discounted pricing

1-(Phenylsulfonyl)pyrrole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(Phenylsulfonyl)pyrrole
CAS 16851-82-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H9NO2S
Formula Weight 207.25
MDL Number MFCD00067739
Synonym N-Benzenesulphonylpyrrole
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC306430010

  SDS
thermo fisher scientific
306430010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 207.25
Physical Form Crystals or Crystalline Powder
Quantity 1g
Color Beige to light brown
Melting Point 86° to 91°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Synonym N-Benzenesulphonylpyrrole
Chemical Name or Material 1-(Phenylsulfonyl)pyrrole
Assay Percent Range 97%
CAS 16851-82-4
Molecular Formula C10H9NO2S
MDL Number MFCD00067739
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.