missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Phenylsulfonyl)-1H-pyrrole, 97%, Maybridge

Chemical Identifiers

CAS 16851-82-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9NO2S
Formula Weight 207.25
MDL Number MFCD00067739
Synonym N-Benzenesulphonylpyrrole
Chemical Name or Material 1-(Phenylsulfonyl)-1H-pyrrole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BTB07001DA SDS Fisher Scientific Company
BTB07001DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

16851-82-4
100.0
C10H9NO2S
MFCD00067739
1-(Phenylsulfonyl)-1H-pyrrole
96.5
97%
207.25
N-Benzenesulphonylpyrrole
97%

Specifications

207.25
100.0
Amber Glass Bottle
96.5
C10H9NO2S
1-(Phenylsulfonyl)-1H-pyrrole
1g
N-Benzenesulphonylpyrrole
97%
MFCD00067739
16851-82-4
97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.