missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenylsulfonyl-1H-pyrazole-4-boronic acid pinacol ester, 95%

$174.26 - $396.32

Chemical Identifiers

CAS 1073372-04-9
Molecular Formula C15H19BN2O4S
Molecular Weight (g/mol) 334.197
MDL Number MFCD09037504
InChI Key NPRHNPBHUVCHKP-UHFFFAOYSA-N
Synonym 1-benzenesulfonyl-1h-pyrazole-4-boronic acid pinacol ester
PubChem CID 44755203
IUPAC Name 1-(benzenesulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)S(=O)(=O)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26954MD View Documents Alfa Aesar™
H26954MD
250mg Each for $174.26
 
AAH2695403 View Documents Alfa Aesar™
H2695403
1g Each for $396.32
 
Specifications

Chemical Identifiers

1073372-04-9
334.197
NPRHNPBHUVCHKP-UHFFFAOYSA-N
44755203
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)S(=O)(=O)C3=CC=CC=C3
C15H19BN2O4S
MFCD09037504
1-benzenesulfonyl-1h-pyrazole-4-boronic acid pinacol ester
1-(benzenesulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole

Specifications

125°C to 130°C
1073372-04-9
MFCD09037504
NPRHNPBHUVCHKP-UHFFFAOYSA-N
1-(benzenesulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
44755203
95%
250mg
C15H19BN2O4S
1-benzenesulfonyl-1h-pyrazole-4-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)S(=O)(=O)C3=CC=CC=C3
334.197
334.2
1-Phenylsulfonyl-1H-pyrazole-4-boronic acid pinacol ester