missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Phenylsulfonyl)-1H-indol-3-ylboronic acid, 97%, May contain varying amounts of anhydri, Maybridge

Manufacturer:  Fisher Scientific Company CC03112EA

 View more versions of this product

Catalog No. CC03112EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-(Phenylsulfonyl)-1H-indol-3-ylboronic acid
10g
129271-98-3
100.0
C14H12BNO4S
1-benzenesulfonyl indol-3-yl boronic acid
B(C1=CN(C2=CC=CC=C12)S(=O)(=O)C3=CC=CC=C3)(O)O
301.123
301.13
Amber Glass Bottle
May contain varying amounts of anhydride
96.5
97%
MFCD02681892
YKTZLHLBQGCFQX-UHFFFAOYSA-N
[1-(benzenesulfonyl)indol-3-yl]boronic acid
2776217
97%

Chemical Identifiers

129271-98-3
301.123
YKTZLHLBQGCFQX-UHFFFAOYSA-N
2776217
B(C1=CN(C2=CC=CC=C12)S(=O)(=O)C3=CC=CC=C3)(O)O
C14H12BNO4S
MFCD02681892
1-benzenesulfonyl indol-3-yl boronic acid
[1-(benzenesulfonyl)indol-3-yl]boronic acid
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents