Please login to your online account to display your discounted pricing

1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid, 97%

Chemical Identifiers

Chemical Name or Material 1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid
Assay Percent Range 97%
Molecular Formula C14H12BNO4S
Formula Weight 301.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC03312DE

 
fisher scientific company
CC03312DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC03312DA

  SDS
fisher scientific company
CC03312DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 301.13
Quantity 5g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C14H12BNO4S
CAS 342404-46-0
Chemical Name or Material 1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid