Learn More
1-Phenylpyrrolidine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0523914
Description
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store cold 2-8°C.
Specifications
1-Phenylpyrrolidine | |
133°C to 134°C (19 mmHg) | |
1.582 | |
25 g | |
1-phenyl-pyrrolidine, n-phenylpyrrolidine, phenylpyrrolidine, pyrrolidine, 1-phenyl, 1-phenyl-pyrrolidin-3-ol, 1-phenylpyrolidine, n-phenyl pyrrolidine, pubchem9192, acmc-1cttc | |
VDQQJMHXZCMNMU-UHFFFAOYSA-N | |
1-phenylpyrrolidine | |
77726 | |
≥98% |
4096-21-3 | |
C10H13N | |
MFCD00015897 | |
117030 | |
Not miscible in water. | |
C1CCN(C1)C1=CC=CC=C1 | |
147.22 | |
147.22 |
Chemical Identifiers
4096-21-3 | |
147.22 | |
VDQQJMHXZCMNMU-UHFFFAOYSA-N | |
77726 | |
C1CCN(C1)C1=CC=CC=C1 |
C10H13N | |
MFCD00015897 | |
1-phenyl-pyrrolidine, n-phenylpyrrolidine, phenylpyrrolidine, pyrrolidine, 1-phenyl, 1-phenyl-pyrrolidin-3-ol, 1-phenylpyrolidine, n-phenyl pyrrolidine, pubchem9192, acmc-1cttc | |
1-phenylpyrrolidine |
Safety and Handling
GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
EINECSNumber : 223-849-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only