Please login to your online account to display your discounted pricing

1-Phenylpyrrole 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  157590250

 View more versions of this product

Catalog No. AC157590250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder and Lumps
Formula Weight 143.19
Quantity 25g
Color Light beige to brown
EINECS Number 211-242-3
TSCA TSCA
Melting Point 58° to 60°C
Beilstein 20, 164
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 1-Phenylpyrrole
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H9N
Linear Formula C6H5(C4H4N)
MDL Number MFCD00005343
CAS 635-90-5
Infrared Spectrum Authentic
CAS Min % 98.5

Specifications

Chemical Name or Material 1-Phenylpyrrole
Name Note 99%
CAS 635-90-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H9N
Linear Formula C6H5(C4H4N)
Formula Weight 143.19
MDL Number MFCD00005343