Please login to your online account to display your discounted pricing

1-Phenylpyrrole 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Phenylpyrrole
Name Note 99%
CAS 635-90-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H9N
Linear Formula C6H5(C4H4N)
Formula Weight 143.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC157590250

  SDS
thermo fisher scientific
157590250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder and Lumps
Formula Weight 143.19
Quantity 25g
Color Light beige to brown
EINECS Number 211-242-3
TSCA TSCA
Melting Point 58° to 60°C
Beilstein 20, 164
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 1-Phenylpyrrole
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H9N
Linear Formula C6H5(C4H4N)
MDL Number MFCD00005343
CAS 635-90-5
Infrared Spectrum Authentic
CAS Min % 98.5