missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenylpiperidine, TCI America™

Manufacturer:  TCI America P21595G

 View more versions of this product

Catalog No. P21595G



Specifications

Specifications

1-Phenylpiperidine
Yellow
4096-20-2
MFCD00129777
C1CCN(CC1)C2=CC=CC=C2
161.248
161.25
≥98.0% (GC,T)
258°C
5g
C11H15N
LLSKXGRDUPMXLC-UHFFFAOYSA-N
1-phenylpiperidine
20038
Clear Liquid at 20°C

Chemical Identifiers

4096-20-2
161.248
LLSKXGRDUPMXLC-UHFFFAOYSA-N
1-phenylpiperidine
C11H15N
MFCD00129777
20038
C1CCN(CC1)C2=CC=CC=C2
Safety and Handling

Safety and Handling

TSCA : Yes

Documents