Please login to your online account to display your discounted pricing

1-Phenylpiperazine, 97%, ACROS Organics™

$109.65 - $109.65

Chemical Identifiers

Chemical Name or Material 1-Phenylpiperazine
CAS 92-54-6
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0 to 104.0%
Assay Percent Range 97%
Molecular Formula C10H14N2
Formula Weight 162.24
MDL Number MFCD00005957
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130821000

  SDS
thermo fisher scientific
130821000
Glass bottle 100g Each for $109.65

AC130820250

  SDS
thermo fisher scientific
AC130820250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 162.24
Quantity 100g
Color Clear colorless to yellow
Assay 96.0 to 104.0%
Density 1.0620g/mL
UN Number 2927
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C10H14N2
Refractive Index 1.5865 to 1.5885 (20°C, 589nm)
Chemical Name or Material 1-Phenylpiperazine
CAS 92-54-6
Assay Percent Range 97%
Health Hazard 1 Danger
MDL Number MFCD00005957
Packaging Glass bottle
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic
Additional Information aniline: < 0.5%
Health Hazard 2 GHS H Statement:
Fatal in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.