missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyloxindole, TCI America™

Manufacturer:  TCI America P176925G

 View more versions of this product

Catalog No. P176925G



Specifications

Specifications

1-Phenyloxindole
122°C
3335-98-6
C14H11NO
1-Phenyl-2-indolinone, 2-Oxo-1-phenylindoline
C1C2=CC=CC=C2N(C1=O)C3=CC=CC=C3
209.248
209.25
Yellow
25g
≥98.0% (GC)
MFCD00234850
OWPNVXATCSXTBK-UHFFFAOYSA-N
1-phenyl-3H-indol-2-one
314787
Crystal-Powder at 20°C

Chemical Identifiers

3335-98-6
209.248
OWPNVXATCSXTBK-UHFFFAOYSA-N
314787
C1C2=CC=CC=C2N(C1=O)C3=CC=CC=C3
C14H11NO
MFCD00234850
1-Phenyl-2-indolinone, 2-Oxo-1-phenylindoline
1-phenyl-3H-indol-2-one
Safety and Handling

Safety and Handling

TSCA : No

Documents