missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Phenylmethyl)-3-pyrrolidinone, 98%, ACROS Organics™

Chemical Identifiers

CAS 775-16-6
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00005342
InChI Key DHGMDHQNUNRMIN-UHFFFAOYSA-N
Synonym 1-benzyl pyrrolidin-3-one, 1-benzyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-phenylmethyl-3-pyrrolidinone, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, n-benzylpyrrolid-3-one, pubchem10007
PubChem CID 69890
IUPAC Name 1-benzylpyrrolidin-3-one
SMILES C1CN(CC1=O)CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC271030010
SDS View Documents
Fisher Scientific Acros
271030010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC271030100
SDS View Documents
Thermo Fisher Scientific
AC271030100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

775-16-6
175.23
DHGMDHQNUNRMIN-UHFFFAOYSA-N
69890
C1CN(CC1=O)CC2=CC=CC=C2
C11H13NO
MFCD00005342
1-benzyl pyrrolidin-3-one, 1-benzyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-phenylmethyl-3-pyrrolidinone, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, n-benzylpyrrolid-3-one, pubchem10007
1-benzylpyrrolidin-3-one

Specifications

>112°C
Glass bottle
109°C (0.1mmHg)
1g
97.5
97.5% min. (GC)
MFCD00005342
1-benzyl pyrrolidin-3-one, 1-benzyl-3-pyrrolidinone, 1-benzyl-3-pyrrolidone, 1-benzyl-pyrrolidin-3-one, 1-phenylmethyl-3-pyrrolidinone, 3-pyrrolidinone, 1-phenylmethyl, benzyl-3-pyrrolidinone, n-benzyl-3-pyrrolidinone, n-benzylpyrrolid-3-one, pubchem10007
C1CN(CC1=O)CC2=CC=CC=C2
175.23
175.23
98%
Authentic
1.5370 to 1.54
White
775-16-6
100.0
C11H13NO
21, V,6, 389
DHGMDHQNUNRMIN-UHFFFAOYSA-N
1-benzylpyrrolidin-3-one
69890
Crystalline Powder, Crystals or Chunks
1-(Phenylmethyl)-3-pyrrolidinone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-274-

Recommended Storage : Product may discolor to yellow-brown on storage

SDS