Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenylimidazole, 99%

$32.26 - $83.93

Chemical Identifiers

Chemical Name or Material 1-Phenylimidazole
CAS 7164-98-9
Molecular Formula C9H8N2
Formula Weight 144.18
MDL Number MFCD00041204
Beilstein 112356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1057903

  SDS
alfa aesar™
L1057903
1g Each for $83.93

AAL10579MD

  SDS
alfa aesar™
L10579MD
250mg Each for $32.26
Description & Specifications

Specifications

Formula Weight 144.18
Chemical Name or Material 1-Phenylimidazole
Assay 99%
Molecular Formula C9H8N2
MDL Number MFCD00041204
CAS 7164-98-9
Beilstein 112356
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Flash Point >110°C (230°F)
Boiling Point 142°C (15mmHg)
Melting Point 13°C
Quantity 1g

Applications
1-phenylimidazole is a ligand for cytochrome P-450 and used as an inhibitor of microsomal oxidation. 1-Phenylimidazole is a suitable reagent used to investigate its effect on the citrulline formation by bovine brain nitric-oxide synthase. 1-Phenylimidazole is reported to be inhibitor of calmodulin-dependent nitric-oxide synthase from bovine brain and GHs pituitary cells.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.