missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenylimidazole, 99%

Manufacturer:  Alfa Aesar™ L10579MD

 View more versions of this product

Catalog No. AAL10579MD



Description

Description

Applications
1-phenylimidazole is a ligand for cytochrome P-450 and used as an inhibitor of microsomal oxidation. 1-Phenylimidazole is a suitable reagent used to investigate its effect on the citrulline formation by bovine brain nitric-oxide synthase. 1-Phenylimidazole is reported to be inhibitor of calmodulin-dependent nitric-oxide synthase from bovine brain and GHs pituitary cells.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

1-Phenylimidazole
142°C (15mmHg)
250mg
C9H8N2
112356
SEULWJSKCVACTH-UHFFFAOYSA-N
1-phenylimidazole
81595
99%
>110°C (230°F)
13°C
7164-98-9
MFCD00041204
1-phenyl-1h-imidazole, phenylimidazole, n-phenylimidazole, 1h-imidazole, 1-phenyl, 1h-imidazole, phenyl, n-phenyl imidazole, pubchem12611, acmc-20aos8, 1-phenylimidazole, 1-phenyl-1h-imidazole #
C1=CC=C(C=C1)N2C=CN=C2
144.177
144.18

Chemical Identifiers

7164-98-9
144.177
SEULWJSKCVACTH-UHFFFAOYSA-N
81595
C1=CC=C(C=C1)N2C=CN=C2
C9H8N2
MFCD00041204
1-phenyl-1h-imidazole, phenylimidazole, n-phenylimidazole, 1h-imidazole, 1-phenyl, 1h-imidazole, phenyl, n-phenyl imidazole, pubchem12611, acmc-20aos8, 1-phenylimidazole, 1-phenyl-1h-imidazole #
1-phenylimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents