Please login to your online account to display your discounted pricing

1-Phenylhexane 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Phenylhexane
Name Note 97%
CAS 1077-16-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12 H18
Linear Formula C6H5(CH2)5CH3
Formula Weight 162.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130661000

  SDS
thermo fisher scientific
130661000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Quantity 100g
Color Clear colorless to light yellow
Beilstein 05, II, 337
Chemical Name or Material 1-Phenylhexane
Molecular Formula C12 H18
Packaging Glass bottle
Refractive Index 1.4855 to 1.4875 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Linear Formula C6H5(CH2)5CH3
MDL Number MFCD00009526
CAS 1077-16-3
Synonym n-Hexylbenzene
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
EINECS Number 214-070-7
TSCA TSCA
Formula Weight 162.27
Density 0.8600g/mL