Please login to your online account to display your discounted pricing

1-Phenylhexane 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  130661000

 View more versions of this product

Catalog No. AC130661000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Quantity 100g
Color Clear colorless to light yellow
Beilstein 05, II, 337
Chemical Name or Material 1-Phenylhexane
Molecular Formula C12 H18
CAS 1077-16-3
MDL Number MFCD00009526
Synonym n-Hexylbenzene
Packaging Glass bottle
Name Note 97%
Infrared Spectrum Authentic
CAS Min % 96.0
Assay Percent Range 97%
CAS Max % 100.0
Linear Formula C6H5(CH2)5CH3
Refractive Index 1.4855 to 1.4875 (20°C, 589nm)
Density 0.8600g/mL
EINECS Number 214-070-7
TSCA TSCA
Formula Weight 162.27

Specifications

Chemical Name or Material 1-Phenylhexane
Name Note 97%
CAS 1077-16-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12 H18
Linear Formula C6H5(CH2)5CH3
Formula Weight 162.27
MDL Number MFCD00009526
Synonym n-Hexylbenzene