Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Phenylethynyl)cyclohexanol, 99%

$177.01 - $177.01

Chemical Identifiers

Chemical Name or Material 1-(Phenylethynyl)cyclohexanol
CAS 20109-09-5
Molecular Formula C14H16O
Formula Weight 200.28
MDL Number MFCD00060883
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2180806

 
alfa aesar™
B2180806
5g Each for $177.01
Description & Specifications

Specifications

Formula Weight 200.28
Chemical Name or Material 1-(Phenylethynyl)cyclohexanol
Assay 99%
Molecular Formula C14H16O
MDL Number MFCD00060883
CAS 20109-09-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 60° to 62°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.