missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenylcyclopropanecarbonitrile, TCI America™

Manufacturer:  TCI America P23235G

 View more versions of this product

Catalog No. P23235G



Specifications

Specifications

1-Phenylcyclopropanecarbonitrile
Yellow
935-44-4
MFCD00001270
ZHFURHRJUWYDKG-UHFFFAOYSA-N
1-phenylcyclopropane-1-carbonitrile
70288
Clear Liquid at 20°C
253°C
5g
C10H9N
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
C1CC1(C#N)C2=CC=CC=C2
143.189
143.19
≥98.0% (GC)

Chemical Identifiers

935-44-4
143.189
ZHFURHRJUWYDKG-UHFFFAOYSA-N
70288
C1CC1(C#N)C2=CC=CC=C2
C10H9N
MFCD00001270
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
1-phenylcyclopropane-1-carbonitrile
Safety and Handling

Safety and Handling

TSCA : No

Documents