Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenylcyclopentanecarboxylic acid, 98%

$36.64 - $136.08

Chemical Identifiers

Chemical Name or Material 1-Phenylcyclopentanecarboxylic acid
CAS 77-55-4
Molecular Formula C12H14O2
Formula Weight 190.24
MDL Number MFCD00001372
Beilstein 1638653
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2550406

 
alfa aesar™
B2550406
5g Each for $36.64

AAB2550414

 
alfa aesar™
B2550414
25g Each for $136.08
Description & Specifications

Specifications

Formula Weight 190.24
Chemical Name or Material 1-Phenylcyclopentanecarboxylic acid
Odor Odorless
Assay 98%
Molecular Formula C12H14O2
MDL Number MFCD00001372
CAS 77-55-4
Beilstein 1638653
Melting Point 158° to 161°C
Quantity 5g