missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenylcyclohexanol, TCI America™

Manufacturer:  TCI America P198825G

 View more versions of this product

Catalog No. P198825G



Specifications

Specifications

1-Phenylcyclohexanol
White-Yellow
25g
C12H16O
1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
C1CCC(CC1)(C2=CC=CC=C2)O
176.259
176.26
≥98.0% (GC)
100°C
63°C
1589-60-2
MFCD00021393
DTTDXHDYTWQDCS-UHFFFAOYSA-N
1-phenylcyclohexan-1-ol
15319
Crystal-Powder at 20°C

Chemical Identifiers

1589-60-2
176.259
DTTDXHDYTWQDCS-UHFFFAOYSA-N
15319
C1CCC(CC1)(C2=CC=CC=C2)O
C12H16O
MFCD00021393
1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
1-phenylcyclohexan-1-ol
Safety and Handling

Safety and Handling

RTECSNumber : GW0699900

TSCA : No

Documents