missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Phenylcyclohexanol 98.0+%, TCI America™
$154.29 - $462.86
Chemical Identifiers
CAS | 1589-60-2 |
---|---|
Molecular Formula | C12H16O |
Molecular Weight (g/mol) | 176.259 |
MDL Number | MFCD00021393 |
InChI Key | DTTDXHDYTWQDCS-UHFFFAOYSA-N |
Synonym | 1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8 |
PubChem CID | 15319 |
IUPAC Name | 1-phenylcyclohexan-1-ol |
SMILES | C1CCC(CC1)(C2=CC=CC=C2)O |
Chemical Identifiers
1589-60-2 | |
176.259 | |
DTTDXHDYTWQDCS-UHFFFAOYSA-N | |
15319 | |
C1CCC(CC1)(C2=CC=CC=C2)O |
C12H16O | |
MFCD00021393 | |
1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8 | |
1-phenylcyclohexan-1-ol |
Specifications
1589-60-2 | |
White-Yellow | |
C12H16O | |
5 g | |
DTTDXHDYTWQDCS-UHFFFAOYSA-N | |
1-phenylcyclohexan-1-ol | |
15319 | |
≥98.0% (GC) | |
1-Phenylcyclohexanol |
63°C | |
100°C | |
MFCD00021393 | |
1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8 | |
C1CCC(CC1)(C2=CC=CC=C2)O | |
176.259 | |
176.26 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : GW0699900
TSCA : No