missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenylcyclohexanol, 97%

$59.53 - $231.86

Chemical Identifiers

CAS 1589-60-2
Molecular Formula C12H16O
Formula Weight 176.26
MDL Number MFCD00021393
Beilstein 1818360
Chemical Name or Material 1-Phenylcyclohexanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2087709 SDS Alfa Aesar™
B2087709
10g Each for $59.53
AAB2087718 SDS Alfa Aesar™
B2087718
50g Each for $231.86
Specifications Description & Specifications

Chemical Identifiers

1589-60-2
176.26
1818360
C12H16O
MFCD00021393
1-Phenylcyclohexanol

Specifications

153°C (20mmHg)
10g
C12H16O
MFCD00021393
1-Phenylcyclohexanol
58° to 62°C
1589-60-2
176.26
1818360
97%

Safety and Handling

RTECSNumber : GW0699900