missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenylcyclohexanecarbonitrile, TCI America™

Manufacturer:  TCI America P225325G

 View more versions of this product

Catalog No. P225325G



Specifications

Specifications

1-Phenylcyclohexanecarbonitrile
Yellow
2201-23-2
MFCD00021281
1-phenylcyclohexanecarbonitrile, 1-phenyl-1-cyclohexanecarbonitrile, cyclohexanecarbonitrile, 1-phenyl, cyanophenylcyclohexane, 1-cyano-1-phenylcyclohexane, 1-phenyl-cyclohexanecarbonitrile, 1-phenylcyclohexane carbonitrile, 1-phenyl-1-cyclohexane-carbonitrile
C1CCC(CC1)(C#N)C2=CC=CC=C2
185.27
185.27
≥97.0% (GC)
141°C
25g
C13H15N
3276
AUXIEQKHXAYAHG-UHFFFAOYSA-N
1-phenylcyclohexane-1-carbonitrile
75148
Clear Liquid at 20°C

Chemical Identifiers

2201-23-2
185.27
AUXIEQKHXAYAHG-UHFFFAOYSA-N
75148
C1CCC(CC1)(C#N)C2=CC=CC=C2
C13H15N
MFCD00021281
1-phenylcyclohexanecarbonitrile, 1-phenyl-1-cyclohexanecarbonitrile, cyclohexanecarbonitrile, 1-phenyl, cyanophenylcyclohexane, 1-cyano-1-phenylcyclohexane, 1-phenyl-cyclohexanecarbonitrile, 1-phenylcyclohexane carbonitrile, 1-phenyl-1-cyclohexane-carbonitrile
1-phenylcyclohexane-1-carbonitrile
Safety and Handling

Safety and Handling

TSCA : No

Documents