missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenylcyclobutanecarbonitrile 95%, ACROS Organics™

Manufacturer:  Acros Organics 154950050

 View more versions of this product

Catalog No. AC154950050



Specifications

Specifications

1-Phenylcyclobutanecarbonitrile
Authentic
1.5313 to 1.5333
5g
14377-68-5
100.0
C11H11N
MFCD00019258
DHIDUDPFTZJPCQ-UHFFFAOYSA-N
1-phenylcyclobutane-1-carbonitrile
84400
Liquid
1.0300g/mL
Glass bottle
Yellow
95%
94.0
95%
C6H5C4H6CN
1-phenylcyclobutanecarbonitrile, cyclobutanecarbonitrile, 1-phenyl, 1-phenyl-cyclobutanecarbonitrile, acmc-1c523, cyclobutanecarbonitrile,1-phenyl, 1-phenyl-1-cyclobutanecarbonitrile, 1-phenyl-1-cyclobutane carbonitrile
C1CC(C1)(C#N)C2=CC=CC=C2
157.216
157.21
95%

Chemical Identifiers

14377-68-5
157.216
DHIDUDPFTZJPCQ-UHFFFAOYSA-N
84400
C1CC(C1)(C#N)C2=CC=CC=C2
C11H11N
MFCD00019258
1-phenylcyclobutanecarbonitrile, cyclobutanecarbonitrile, 1-phenyl, 1-phenyl-cyclobutanecarbonitrile, acmc-1c523, cyclobutanecarbonitrile,1-phenyl, 1-phenyl-1-cyclobutanecarbonitrile, 1-phenyl-1-cyclobutane carbonitrile
1-phenylcyclobutane-1-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-351-9

TSCA : TSCA

SDS Documents