Please login to your online account to display your discounted pricing

1-Phenyl-5-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Phenyl-5-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride
CAS 175137-14-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H6ClF3N2O
Formula Weight 274.63
MDL Number MFCD00068139
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK01494DA

 
fisher scientific company
GK01494DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Assay 97%
Formula Weight 274.63
CAS Max % 100.0
Chemical Name or Material 1-Phenyl-5-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride
Assay Percent Range 97%
Molecular Formula C11H6ClF3N2O
MDL Number MFCD00068139
CAS 175137-14-1
Packaging Amber Glass Bottle
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.