missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenyl-3-trimethylsilyl-2-propyn-1-ol, 97%

Manufacturer:  Alfa Aesar™ H5342614

 View more versions of this product

Catalog No. AAH5342614



Specifications

Specifications

1-Phenyl-3-trimethylsilyl-2-propyn-1-ol
C12H16OSi
1-phenyl-3-trimethylsilyl-2-propyn-1-ol, 1-phenyl-3-trimethylsilyl prop-2-yn-1-ol, benzenemethanol, a-2-trimethylsilyl ethynyl, acmc-20ln8h, alpha-trimethylsilyl ethynylbenzyl alcohol, 4,4-dimethyl-1-phenyl-4-silapent-2-yn-1-ol, alpha-trimethylsilyl ethynyl-benzenemethanol, alpha-trimethylsilyl-ethynyl-benzenemethanol, alpha-trimethylsilyl ethynyl-benzene-methanol, 1-phenyl-3-1,1,1-trimethylsilyl-2-propyn-1-ol
C[Si](C)(C)C#CC(C1=CC=CC=C1)O
204.344
204.34
25g
89530-34-7
MFCD05864336
RVTDALNDYLDVMN-UHFFFAOYSA-N
1-phenyl-3-trimethylsilylprop-2-yn-1-ol
2760416
97%
110°C (230°F)

Chemical Identifiers

89530-34-7
204.344
RVTDALNDYLDVMN-UHFFFAOYSA-N
2760416
C[Si](C)(C)C#CC(C1=CC=CC=C1)O
C12H16OSi
MFCD05864336
1-phenyl-3-trimethylsilyl-2-propyn-1-ol, 1-phenyl-3-trimethylsilyl prop-2-yn-1-ol, benzenemethanol, a-2-trimethylsilyl ethynyl, acmc-20ln8h, alpha-trimethylsilyl ethynylbenzyl alcohol, 4,4-dimethyl-1-phenyl-4-silapent-2-yn-1-ol, alpha-trimethylsilyl ethynyl-benzenemethanol, alpha-trimethylsilyl-ethynyl-benzenemethanol, alpha-trimethylsilyl ethynyl-benzene-methanol, 1-phenyl-3-1,1,1-trimethylsilyl-2-propyn-1-ol
1-phenyl-3-trimethylsilylprop-2-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

SDS Documents