missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenyl-3-pyrazolidinone, 97%

Manufacturer:  Alfa Aesar™ A1208930

 View more versions of this product

Catalog No. AAA1208930



Description

Description

Applications
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.

Solubility
Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

1-Phenyl-3-pyrazolidinone
250g
C9H10N2O
UN2811
14,7309
CMCWWLVWPDLCRM-UHFFFAOYSA-N
1-phenylpyrazolidin-3-one
7090
97%
120°C to 123°C
92-43-3
MFCD00003094
131856
phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
C1CN(NC1=O)C2=CC=CC=C2
162.192
162.19

Chemical Identifiers

92-43-3
162.192
CMCWWLVWPDLCRM-UHFFFAOYSA-N
7090
C1CN(NC1=O)C2=CC=CC=C2
C9H10N2O
MFCD00003094
phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
1-phenylpyrazolidin-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : UQ8750000

TSCA : TSCA

SDS Documents