missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenyl-2-trimethylsilylacetylene, 99%

Manufacturer:  Alfa Aesar™ H5586614

 View more versions of this product

Catalog No. AAH5586614



Description

Description

Applications
1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

Solubility
Sparingly Soluble (6.1E-3 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications

Specifications

1-Phenyl-2-trimethylsilylacetylene
74°C (165°F)
87°C to 88°C (9mmHg)
25g
C16H40HfO4
MFCD00054860
hafnium; 2-methylpropan-2-ol
YZABPPRMRYHJGL-UHFFFAOYSA-N
hafnium;2-methylpropan-2-ol
25199745
Liquid
0.886 g/mL
1.528
Colorless
2170-06-1
C6H5C≡CSi(CH3)3
3088678
Sparingly Soluble (6.1E-3 g/L) (25°C)
CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Hf]
474.98
174.32
99%

Chemical Identifiers

2170-06-1
474.98
YZABPPRMRYHJGL-UHFFFAOYSA-N
25199745
CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Hf]
C16H40HfO4
MFCD00054860
hafnium; 2-methylpropan-2-ol
hafnium;2-methylpropan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents