Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenyl-2-hexanone, 95%

$37.57 - $571.07

Chemical Identifiers

Chemical Name or Material 1-Phenyl-2-hexanone
CAS 25870-62-6
Molecular Formula C12H16O
Formula Weight 176.26
MDL Number MFCD00027240
Beilstein 2044423
Synonym Benzyl n-butyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1094403

  SDS
alfa aesar™
A1094403
1g Each for $37.57

AAA1094406

  SDS
alfa aesar™
A1094406
5g Each for $137.31

AAA1094414

  SDS
alfa aesar™
A1094414
25g Each for $571.07
Description & Specifications

Specifications

Formula Weight 176.26
Chemical Name or Material 1-Phenyl-2-hexanone
Assay 95%
Molecular Formula C12H16O
MDL Number MFCD00027240
CAS 25870-62-6
Beilstein 2044423
Synonym Benzyl n-butyl ketone
Flash Point >110°C (230°F)
Boiling Point 153°C (20mmHg)
Quantity 1g