Please login to your online account to display your discounted pricing

1-Phenyl-2-butanone, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC159880050

 View more versions of this product

Catalog No. AC159880050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 148.2
Quantity 5g
Color Clear colorless
Density 0.99g/mL
CAS Max % 100.0
Synonym Benzyl ethyleketone
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-Phenyl-2-butanone
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H12O
CAS 1007-32-5
CAS Min % 97.5
Linear Formula C6H5CH2COC2H5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-Phenyl-2-butanone
CAS 1007-32-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O
Linear Formula C6H5CH2COC2H5
Formula Weight 148.2
Synonym Benzyl ethyleketone
Accessories

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.