missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-2-butanone, 98%, ACROS Organics™

Chemical Identifiers

CAS 1007-32-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10H12O
Linear Formula C6H5CH2COC2H5
Formula Weight 148.2
Synonym Benzyl ethyleketone
Chemical Name or Material 1-Phenyl-2-butanone
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC159880050 Thermo Fisher Scientific
AC159880050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1007-32-5
100.0
C10H12O
148.2
1-Phenyl-2-butanone
97.5
98%
C6H5CH2COC2H5
Benzyl ethyleketone
≥97.5%

Specifications

0.99g/mL
5g
97.5
Authentic
Liquid
C10H12O
148.2
1-Phenyl-2-butanone
Clear colorless
1007-32-5
100.0
Glass bottle
98%
C6H5CH2COC2H5
Benzyl ethyleketone
≥97.5%