Please login to your online account to display your discounted pricing

1-Phenyl-1H-pyrazole-5-carbaldehyde, 95+%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC24504CB

 View more versions of this product

Catalog No. CC24504CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 172.19
Quantity 250mg
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H8N2O
CAS 132274-70-5
Packaging Amber Glass Bottle
Chemical Name or Material 1-Phenyl-1H-pyrazole-5-carbaldehyde

Specifications

Chemical Name or Material 1-Phenyl-1H-pyrazole-5-carbaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H8N2O
Formula Weight 172.19