missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%

$59.23 - $191.22

Chemical Identifiers

CAS 54605-72-0
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD02179568
InChI Key PHVRLPFVPVKYOI-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole-4-carbaldehyde, 1-phenyl-1h-pyrazole-4-carboxaldehyde, 1-phenylpyrazole-4-carboxaldehyde, pubchem13271, acmc-1awxu, phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,1-phenyl
PubChem CID 952089
IUPAC Name 1-phenylpyrazole-4-carbaldehyde
SMILES C1=CC=C(C=C1)N2C=C(C=N2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63695MD View Documents Alfa Aesar™
H63695MD
250mg Each for $59.23
 
AAH6369503 View Documents Alfa Aesar™
H6369503
1g Each for $191.22
 
Specifications

Chemical Identifiers

54605-72-0
172.187
PHVRLPFVPVKYOI-UHFFFAOYSA-N
952089
C1=CC=C(C=C1)N2C=C(C=N2)C=O
C10H8N2O
MFCD02179568
1-phenyl-1h-pyrazole-4-carbaldehyde, 1-phenyl-1h-pyrazole-4-carboxaldehyde, 1-phenylpyrazole-4-carboxaldehyde, pubchem13271, acmc-1awxu, phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,1-phenyl
1-phenylpyrazole-4-carbaldehyde

Specifications

Moisture & Light Sensitive
84°C to 88°C
54605-72-0
MFCD02179568
PHVRLPFVPVKYOI-UHFFFAOYSA-N
1-phenylpyrazole-4-carbaldehyde
952089
Solid
1-Phenyl-1H-pyrazole-4-carboxaldehyde
Yellow
250mg
C10H8N2O
1-phenyl-1h-pyrazole-4-carbaldehyde, 1-phenyl-1h-pyrazole-4-carboxaldehyde, 1-phenylpyrazole-4-carboxaldehyde, pubchem13271, acmc-1awxu, phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,1-phenyl
C1=CC=C(C=C1)N2C=C(C=N2)C=O
172.187
172.18
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning