Learn More
1-Phenyl-1-trimethylsiloxyethylene, 95%, Thermo Scientific Chemicals
$50.44 - $170.44
Chemical Identifiers
CAS | 13735-81-4 |
---|---|
Molecular Formula | C11H16OSi |
Molecular Weight (g/mol) | 192.333 |
MDL Number | MFCD00008582 |
InChI Key | AFFPCIMDERUIST-UHFFFAOYSA-N |
Synonym | 1-phenyl-1-trimethylsiloxyethylene, trimethyl 1-phenylvinyl oxy silane, 1-phenyl-1-trimethylsilyloxyethylene, 1-phenyl-1-trimethylsiloxy ethylene, 1-phenyl-1-trimethylsilyloxy ethylene, benzene, 1-trimethylsilyl oxy ethenyl, silane, trimethyl 1-phenylethenyl oxy, acmc-209ccf |
PubChem CID | 117406 |
IUPAC Name | trimethyl(1-phenylethenoxy)silane |
SMILES | C[Si](C)(C)OC(=C)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0530803
|
Thermo Scientific Chemicals
L0530803 |
1 g |
Each for $50.44
|
|
|||||
AAL0530806
|
Thermo Scientific Chemicals
L0530806 |
5 g |
Each for $170.44
|
|
|||||
Description
1-Phenyl-1-trimethylsiloxyethylene has been used in the synthesis of β-amino ketones in water via Mannich-type reaction and cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction. It is also used as chemical additives and intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Phenyl-1-trimethylsiloxyethylene has been used in the synthesis of β-amino ketones in water via Mannich-type reaction and cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction. It is also used as chemical additives and intermediates.
Solubility
Reacts with water.
Notes
Store at 4°C. Moisture Sensitive. Store under inert gas. Incompatible materials are Oxidizing agents.
Chemical Identifiers
13735-81-4 | |
192.333 | |
AFFPCIMDERUIST-UHFFFAOYSA-N | |
117406 | |
C[Si](C)(C)OC(=C)C1=CC=CC=C1 |
C11H16OSi | |
MFCD00008582 | |
1-phenyl-1-trimethylsiloxyethylene, trimethyl 1-phenylvinyl oxy silane, 1-phenyl-1-trimethylsilyloxyethylene, 1-phenyl-1-trimethylsiloxy ethylene, 1-phenyl-1-trimethylsilyloxy ethylene, benzene, 1-trimethylsilyl oxy ethenyl, silane, trimethyl 1-phenylethenyl oxy, acmc-209ccf | |
trimethyl(1-phenylethenoxy)silane |
Specifications
53°C to 54°C (1mmHg) | |
1 g | |
MFCD00008582 | |
78°C (172°F) | |
Reacts with water. | |
C[Si](C)(C)OC(=C)C1=CC=CC=C1 | |
192.333 | |
192.34 | |
1.502 | |
0.938 |
13735-81-4 | |
C11H16OSi | |
1306914 | |
1-phenyl-1-trimethylsiloxyethylene, trimethyl 1-phenylvinyl oxy silane, 1-phenyl-1-trimethylsilyloxyethylene, 1-phenyl-1-trimethylsiloxy ethylene, 1-phenyl-1-trimethylsilyloxy ethylene, benzene, 1-trimethylsilyl oxy ethenyl, silane, trimethyl 1-phenylethenyl oxy, acmc-209ccf | |
AFFPCIMDERUIST-UHFFFAOYSA-N | |
trimethyl(1-phenylethenoxy)silane | |
117406 | |
97% | |
Moisture sensitive | |
1-Phenyl-1-trimethylsiloxyethylene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 237-308-1
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only