Please login to your online account to display your discounted pricing

1-Phenyl-1-propanol 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: fisher scientific acros  150032500

 View more versions of this product

Catalog No. AC150032500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 136.19
Quantity 250g
Color Clear colorless
EINECS Number 202-256-0
TSCA TSCA
RTECS Number DO5470000
Density 0.9900g/mL
CAS Max % 100.0
Synonym α-Ethylbenzyl alcohol
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-Phenyl-1-propanol
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula C2H5CH(C6H5)OH
MDL Number MFCD00004564
CAS 93-54-9
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Packaging Glass bottle
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)

Specifications

Chemical Name or Material 1-Phenyl-1-propanol
Name Note 99%
CAS 93-54-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula C2H5CH(C6H5)OH
Formula Weight 136.19
MDL Number MFCD00004564
Synonym α-Ethylbenzyl alcohol
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents