Please login to your online account to display your discounted pricing

1-Phenyl-1-propanol 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Phenyl-1-propanol
Name Note 99%
CAS 93-54-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula C2H5CH(C6H5)OH
Formula Weight 136.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC150032500

  SDS
fisher scientific acros
150032500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 136.19
Quantity 250g
Color Clear colorless
TSCA TSCA
EINECS Number 202-256-0
RTECS Number DO5470000
Density 0.9900g/mL
CAS Max % 100.0
Synonym α-Ethylbenzyl alcohol
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
CAS 93-54-9
Chemical Name or Material 1-Phenyl-1-propanol
Linear Formula C2H5CH(C6H5)OH
Molecular Formula C9H12O
Name Note 99%
MDL Number MFCD00004564
Assay Percent Range 99%