Please login to your online account to display your discounted pricing

1-Phenyl-1-propanol 99%, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC150030010

 View more versions of this product

Catalog No. AC150030010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 136.19
Physical Form Liquid
Assay ≥98.5%
Quantity 1kg
Color Clear colorless
TSCA TSCA
EINECS Number 202-256-0
RTECS Number DO5470000
Density 0.9900g/mL
CAS Max % 100.0
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym α-Ethylbenzyl alcohol
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Name Note 99%
MDL Number MFCD00004564
Linear Formula C2H5CH(C6H5)OH
CAS 93-54-9
Assay Percent Range 99%
Chemical Name or Material 1-Phenyl-1-propanol
Molecular Formula C9H12O

Specifications

Chemical Name or Material 1-Phenyl-1-propanol
Name Note 99%
CAS 93-54-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula C2H5CH(C6H5)OH
Formula Weight 136.19
MDL Number MFCD00004564
Synonym α-Ethylbenzyl alcohol
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents