missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenyl-1-pentyn-3-ol, 97%

Manufacturer:  Alfa Aesar™ H5341614

 View more versions of this product

Catalog No. AAH5341614



Description

Description

Applications
Reactant in the heterogeneously catalyzed etherification of glycerol, also in fritsch-wiechell rearrangement of 1-chlorovinyl sulfoxides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

1-Phenyl-1-pentyn-3-ol
25g
C11H12O
1-phenyl-1-pentyn-3-ol, 1-pentyn-3-ol,1-phenyl, 1-pentyn-3-ol, 1-phenyl-, s, acmc-20mqra, 1-pentyn-3-ol, 1-phenyl, 1-phenyl-1-pentyn-3-ol
CCC(C#CC1=CC=CC=C1)O
160.216
160.22
>110°C (230°F)
27975-78-6
MFCD00041589
QWCMSASONHVIHV-UHFFFAOYSA-N
1-phenylpent-1-yn-3-ol
11789508
97%

Chemical Identifiers

27975-78-6
160.216
QWCMSASONHVIHV-UHFFFAOYSA-N
11789508
CCC(C#CC1=CC=CC=C1)O
C11H12O
MFCD00041589
1-phenyl-1-pentyn-3-ol, 1-pentyn-3-ol,1-phenyl, 1-pentyn-3-ol, 1-phenyl-, s, acmc-20mqra, 1-pentyn-3-ol, 1-phenyl, 1-phenyl-1-pentyn-3-ol
1-phenylpent-1-yn-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents