missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1-hexanol, TCI America™

Manufacturer:  TCI America P168325G

 View more versions of this product

Catalog No. P168325G



Specifications

Specifications

1-Phenyl-1-hexanol
Yellow
4471-05-0
MFCD02258728
SVCRDVHXRDRHCP-UHFFFAOYSA-N
1-phenylhexan-1-ol
221292
Clear Liquid at 20°C
172°C
25g
C12H18O
alpha-Pentylbenzyl Alcohol
CCCCCC(C1=CC=CC=C1)O
178.275
178.28
≥98.0% (GC)

Chemical Identifiers

4471-05-0
178.275
SVCRDVHXRDRHCP-UHFFFAOYSA-N
221292
CCCCCC(C1=CC=CC=C1)O
C12H18O
MFCD02258728
alpha-Pentylbenzyl Alcohol
1-phenylhexan-1-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents