missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1-cyclopropanecarboxylic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 170680100

 View more versions of this product

Catalog No. AC170680100


 

Specifications Description & Specifications

Specifications

1-Phenyl-1-cyclopropanecarboxylic acid
84° to 87°C
97%
96.0
Authentic
Crystals, Crystalline Powder or Chunks
97%
C6H5C3H4CO2H
MFCD00001288
White to beige
10g
6120-95-2
100.0
Glass bottle
≥96.0%
C10H10O2
162.19

Chemical Identifiers

6120-95-2
100.0
C10H10O2
162.19
1-Phenyl-1-cyclopropanecarboxylic acid, 97%
96.0
97%
C6H5C3H4CO2H
MFCD00001288
≥96.0%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 228-090-9

TSCA : TSCA