missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1-cyclopropanecarbonitrile, 97%, ACROS Organics™

Manufacturer:  Acros Organics 170590500

 View more versions of this product

Catalog No. AC170590500



Specifications

Specifications

1-Phenyl-1-cyclopropanecarbonitrile
>110°C
Glass bottle
133.0°C to 137.0°C (30.0mmHg)
50g
935-44-4
100.0
C10H9N
MFCD00001270
ZHFURHRJUWYDKG-UHFFFAOYSA-N
1-phenylcyclopropane-1-carbonitrile
70288
Liquid
1.0000g/mL
Authentic
1.5370 to 1.5400
Colorless to Yellow
97%
96.0
97%
C6H5C3H4CN
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
C1CC1(C#N)C2=CC=CC=C2
143.189
143.19
97%

Chemical Identifiers

935-44-4
143.189
ZHFURHRJUWYDKG-UHFFFAOYSA-N
70288
C1CC1(C#N)C2=CC=CC=C2
C10H9N
MFCD00001270
1-phenylcyclopropanecarbonitrile, 1-phenyl-1-cyclopropanecarbonitrile, cyclopropanecarbonitrile, 1-phenyl, 1-cyano-1-phenylcyclopropane, 1-phenyl-cyclopropanecarbonitrile, 1-phenylcyclopropyl cyanide, phenylcyclopropanenitrile, 1-phenylcyclopropanenitrile, phenylcyclopropanecarbonitrile, 1-phenylcyclopropane carbonitrile
1-phenylcyclopropane-1-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-304-5

TSCA : TSCA

SDS Documents