Please login to your online account to display your discounted pricing

1-phenyl-1-cyclopentanecarbonitrile, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC136100100

 View more versions of this product

Catalog No. AC136100100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 171.24
Physical Form Liquid
Quantity 10g
Color Clear light yellow
Density 1.0300g/mL
Beilstein 09, IV, 2115
CAS Max % 100.0
Chemical Name or Material 1-Phenyl-1-cyclopentanecarbonitrile
Refractive Index 1.5325 to 1.5345 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C12H13N
Linear Formula C6H5C5H8CN
MDL Number MFCD00001358
CAS 77-57-6
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 1-Phenyl-1-cyclopentanecarbonitrile
CAS 77-57-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H13N
Linear Formula C6H5C5H8CN
Formula Weight 171.24
MDL Number MFCD00001358
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

SDS & Safety Documents