Please login to your online account to display your discounted pricing

1-Phenyl-1-cyclohexene 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Phenyl-1-cyclohexene
Name Note 97%
CAS 771-98-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H14
Linear Formula C6H5C6H9
Formula Weight 158.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC130450250

  SDS
fisher scientific acros
130450250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC130450050

  SDS
fisher scientific acros
130450050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 158.24
Quantity 25g
Color Clear colorless to light yellow
Beilstein 05, II, 419
RTECS Number GW6600000
TSCA TSCA
EINECS Number 212-242-6
Density 0.99g/mL
CAS Min % 94.0
CAS Max % 100.0
Refractive Index 1.5680 to 1.5700 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5C6H9
CAS 771-98-2
MDL Number MFCD00001542
Assay Percent Range 95%
Chemical Name or Material 1-Phenyl-1-cyclohexene
Name Note 97%
Molecular Formula C12H14