Please login to your online account to display your discounted pricing

1-Phenyl-1-butyne, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Phenyl-1-butyne
CAS 622-76-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10
Linear Formula C6H5C2CCH2CH3
Formula Weight 130.19
MDL Number MFCD00039945
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC384070250

  SDS
thermo fisher scientific
AC384070250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC384070050

  SDS
fisher scientific acros
AC384070050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 130.19
Physical Form Liquid
Quantity 25g
Color Clear colorless
Density 0.916g/mL
CAS Max % 100.0
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
Linear Formula C6H5C2CCH2CH3
Molecular Formula C10H10
Chemical Name or Material 1-Phenyl-1-butyne
Assay Percent Range 98%
CAS 622-76-4
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00039945
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.