missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1-butyne, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC384070250

 View more versions of this product

Catalog No. AC384070250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1-Phenyl-1-butyne
Clear colorless
622-76-4
100.0
Glass bottle
Liquid
98%
C6H5C2CCH2CH3
MFCD00039945
0.916g/mL
25g
97.5
Authentic
1.5500 to 1.5520 (20°C, 589nm)
≥97.5%
C10H10
130.19

Chemical Identifiers

622-76-4
100.0
C10H10
130.19
1-Phenyl-1-butyne
97.5
98%
C6H5C2CCH2CH3
MFCD00039945
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.