missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Phenyl-1,4-pentanedione, 96%

Manufacturer:  Alfa Aesar™ L1315106

 View more versions of this product

Catalog No. AAL1315106



Description

Description

Applications
1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1-Phenyl-1,4-pentanedione
5g
C11H12O2
908230
RBLXWIPBPPVLPU-UHFFFAOYSA-N
1-phenylpentane-1,4-dione
136371
96%
158°C to 162°C (12mmHg)
583-05-1
MFCD00092889
1-phenyl-1,4-pentanedione, 1-phenyl-pentane-1,4-dione, 1,4-pentanedione, 1-phenyl, acmc-20aosh, 1-phenyl-1,4-pentandione, 5-phenyl-2,5-pentanedione, benzil-related compound, 41, 1-phenyl pentane-1,4-dione, 1-phenyl-pentane-1,4-dione
CC(=O)CCC(=O)C1=CC=CC=C1
176.215
176.22

Chemical Identifiers

583-05-1
176.215
RBLXWIPBPPVLPU-UHFFFAOYSA-N
136371
CC(=O)CCC(=O)C1=CC=CC=C1
C11H12O2
MFCD00092889
1-phenyl-1,4-pentanedione, 1-phenyl-pentane-1,4-dione, 1,4-pentanedione, 1-phenyl, acmc-20aosh, 1-phenyl-1,4-pentandione, 5-phenyl-2,5-pentanedione, benzil-related compound, 41, 1-phenyl pentane-1,4-dione, 1-phenyl-pentane-1,4-dione
1-phenylpentane-1,4-dione
Documents