missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1,2-ethanediol, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 13059-0050

 View more versions of this product

Catalog No. AC130590050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
138.17
White to light beige
06, 907
96.0
Authentic
C8H10O2
93-56-1
Glass bottle
Styrene glycol
Crystalline Powder or Flakes
5g
15, 8991
61° to 67°C
100.0
97%
1-Phenyl-1, 2-ethanediol
MFCD00003546
HOCH2CH(C6H5)OH

Chemical Identifiers

93-56-1
100.0
C8H10O2
138.17
Styrene glycol
≥96.0%
96.0
97%
HOCH2CH(C6H5)OH
MFCD00003546
1-Phenyl-1, 2-ethanediol